missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ Pro HT NoSpin Plasmid MiniPrep Kit

Product Code. p-200109711 Shop All Applied Biosystems Products
Click to view available options
No. of Reactions:
1152 Reactions
2304 Reactions
384 Reactions
96 Reactions
Unit Size:
1152 reactions
2304 reactions
384 reactions
96 reactions
This item is not returnable. View return policy

Product Code. 30057386

Brand: Applied Biosystems™ A58309

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

The MagMAX Pro HT NoSpin Plasmid Miniprep Kit enables scalable, ultra-rapid purification of high-quality, transfection-grade plasmid DNA (pDNA) from bacterial cultures. Using magnetic-bead technology, it simplifies workflows by eliminating centrifugation and vacuum filtration bottlenecks.

The Applied Biosystems MagMAX Pro HT NoSpin Plasmid Miniprep Kit provides a scalable and ultra-rapid approach for purifying high quality, transfection-grade plasmid DNA (pDNA) directly from bacterial culture. Utilizing advanced magnetic-bead technology, the MagMAX Pro HT NoSpin kit drastically simplifies and streamlines plasmid purification workflows by eliminating centrifugation and vacuum filtration bottlenecks.

MagMAX Pro HT NoSpin Plasmid Miniprep Kit features include:

  • Bead-based—utilizes dual para-magnetic beads for plasmid isolation without time-intensive processing steps (centrifugation, vacuum filtration)
  • Automation enabled—turn-key, automation-friendly protocols and scripts, allowing 96 samples to be processed in under 35 minutes on KingFisher Apex and KingFisher Flex systems
  • Fast plasmid DNA isolation—35 minutes from start to completion and ready for downstream applications
  • Wide range of compatibility—typical E. coli cultures, common medias, and range of plasmids all acceptable
  • High-quality pDNA yields of up to 20 μg, ready for downstream applications
  • Ready for transfection—isolated pDNA with very low endotoxin levels < 1 EU/μg

Specifically designed for high-throughput plasmid isolation, the MagMAX Pro HT NoSpin technology is automation friendly and suited for use with KingFisher Apex, KingFisher Flex, and KingFisher Presto purification systems, as well as integrated liquid handling robots.

The MagMAX Pro HT purification chemistry is both effective and ecofriendly. Kits include a novel endotoxin reduction buffer to remove pyrogenic contaminates, and buffers are free of ethanol and chaotropic salts.

pDNA purified with the MagMAX Pro HT NoSpin kit is suitable for a wide array of downstream applications, such as transfection, cloning, enzymatic restriction, in vitro transcription, and sequencing.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C BactBind Beads, PlasmidPure Beads, Endotoxin Reduction Buffer
15°C to 25°C RNAse A, Resuspension Buffer, Lysis Buffer, Precipitation Buffer, Wash 1 Buffer, Wash 2 Buffer, Elution Buffer
Format 96 Deep-well Plate
Isolation Technology Magnetic Bead
Scale Mini
Sample Type Bacteria, E. coli
Elution Volume 100 μL
For Use With (Equipment) KingFisher Apex, Flex and Presto Processors with 96 Deep Well Heads, Liquid Handling Robotic Systems
For Use With (Application) Cloning, Transfection, Protein Expression, Protein Production, mRNA production via In Vitro Transcription, Gene Editing, Sequencing, PCR, Nucleic acid labeling, PCR
Green Features Energy-efficient design, reduced packaging, and elimination of volatile organic solvents
No. of Reactions 96 Reactions
Nucleic Acid Type Plasmid DNA (pDNA)
Plate Compatibility KingFisher 96 Deep Well Plates with or without barcodes
Product Type Automated, pDNA extraction kit
Quantity 1 Ea.
Purity Transfection Grade
Sample Volume Bacterial Culture, 1 mL per well
Test Time ~ 32 min. for 96 samples, 1 KingFisher 96 DWP
Yield Up to 20 μg pDNA using high copy plasmid
Size Distribution 5-12kb
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.