missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ Pure Bind Beads

Product Code. 17912101 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
5 mL
50 mL
250 mL
Unit Size:
50mL
5mL
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17912101 50 mL 50mL
17992061 5 mL 5mL
17992151 250 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17912101 Supplier Applied Biosystems™ Supplier No. A58522
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MagMAX Pure Bind Beads are ready-to-use magnetic beads that provide optimized size selection and PCR cleanup, removing small fragments such as dNTPs, salts, primers, and primer-dimers.

MagMAX Pure Bind Beads are ready-to-use magnetic beads that provide optimized PCR cleanup, removing small fragments such as dNTPs, salts, primers, and primer-dimers. The magnetic bead technology allows for consistent binding capacity and highly reproducible results. The cleanup protocol is optimized for DNA fragments larger than 90 bp.

MagMAX Pure Bind Beads can seamlessly integrate into established workflows using magnetic beads for PCR cleanup. They can be used to clean up impurities throughout next-generation sequencing workflows.

Features of MagMAX Pure Bind Beads include:

  • >90% recovery of DNA fragments larger than 90 bp
  • Flexibility of starting sample volumes with the ability to scale
  • Consistent results
  • Compatible with manual and automated processes
  • Room temperature storage

The use of MagMAX Pure Bind Beads does not require multiple bottles or long protocols. This product consists of just one bottle containing the optimized magnetic bead formulation, allowing a seamless transition into established workflows.

Sustainability
Additionally, MagMAX Pure Bind Beads are considered a more sustainable/greener product that can be shipped and stored at room temperature, allowing a grab-and-go effect to cut down on lab time. With the use of magnetic beads, the protocol can be completed manually or on automated systems, like KingFisher instruments.

Increase efficiency with compatible workflow products

KingFisher automated purification systems:
MagMAX Pure Bind Beads have been optimized for use on KingFisher automated purification systems. Choose from the KingFisher Apex (Cat. No. 5400910) and KingFisher Flex (Cat. No. 5400610) systems to streamline your DNA cleanup. Reduce sample processing hands-on time while achieving excellent PCR cleanup quality and reproducible results. Users can process 60 μL of PCR sample volume in high-throughput KingFisher 96 deep-well plates. BindIx and BindIt software scripts for KingFisher Apex and Flex systems can be downloaded below.

Streptavidin beads for target sequence enrichment:
For NGS library construction within NGS workflows, MagMAX Pure Bind Beads can be used in conjunction with Dynabeads Streptavidin for Target Enrichment (Cat. No. 65605D) to increase library concentrations and enhance sensitivity.

TRUSTED_SUSTAINABILITY

Specifications

GC-Rich PCR Performance High
For Use With (Application) NGS Cleanup, PCR Cleanup, PCR Purification, DNA Purification, Sequencing
Quantity 50 mL
Test Time 20 min.
Form Beads in Suspension
Can I use MagMax Pure Bind Beads for cleanup of cycle sequencing reactions in Sanger Sequencing?

We do not recommend using MagMax Pure Bind Beads for cleanup of cycle sequencing reactions. Fragments below 90 bp will be lost, so a large part of the beginning of a sequence can not be sequenced following cleanup using MagMax Pure Bind beads.

How long can MagMAX Pure Bind beads be stored at ambient temperatures?

MagMAX Pure Bind beads are stable at ambient temperatures for 18 months post-production.

Are MagMAX Pure Bind beads compatible with automated workflows?

Yes, MagMAX Pure Bind beads can be used with KingFisher Apex, KingFisher Flex, and KingFisher Duo instruments.

Can I perform size selection with MagMAX Pure Bind beads?

Yes, size selection can be performed by adjusting the ratio of MagMAX Pure Bind beads and sample.

What is the smallest size fragment that can be recovered using MagMAX Pure Bind beads?

MagMAX Pure Bind beads are successful in recovering fragments down to 90 bp in size.

What is the expected recovery for MagMAX Pure Bind beads?

MagMAX Pure Bind beads are designed for purification of PCR products that are 90 bp or larger in size. The recovery ranges from 80-90%.

Can I use MagMAX Pure Bind beads to purify RNA?

MagMAX Pure Bind beads have been designed to bind to DNA and are not manufactured under RNase-free conditions. Therefore, the product has not been tested to be RNase-free and we do not recommend using it for RNA cleanup. We offer several RNA purification kits to choose from to suit your specific needs.

If my MagMAX Pure Bind beads were stored at 4 degrees C, can they still be used?

Yes, MagMAX Pure Bind beads can be stored at 4 degrees C and used if they are brought up to room temperature and mixed well before use.

Can I store MagMAX Pure Bind beads at ambient temperatures?

Yes, MagMAX Pure Bind beads were developed to be stored at ambient temperatures to provide a more sustainable solution to our customers and to allow for a grab-and-go workflow.


Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.