missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ Viral/Pathogen Ultra Nucleic Acid Isolation Kit

Product Code. 16370022
Change view
Click to view available options
Quantity:
100 preps
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16370022 100 preps Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16370022 Supplier Applied Biosystems™ Supplier No. A42356
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The MagMAX Viral/Pathogen Ultra Nucleic Acid Isolation Kit contains the same reagents as the standard MagMAX Viral/Pathogen Nucleic Acid Isolation Kit and so, like that kit, recovers nucleic acid from viral particles and gram negative bacteria.

  • Includes a proprietary enzyme mix that enables lysis and nucleic acid recovery from the toughest pathogens, such as gram-positive bacteria, yeast, and fungi
  • With this enzyme mix, used in conjunction with optimized reagents, the MagMAX Viral/Pathogen Ultra kit makes it possible to achieve comprehensive pathogen lysis and recovery of RNA and DNA
  • Utilizes MagMAX magnetic-bead technology, ensuring reproducible recovery of high-quality nucleic acid compatible with a broad range of applications, including real-time PCR, digital PCR, and next-generation sequencing
  • Automation-ready protocols designed for the KingFisher Flex and KingFisher Duo Prime systems
  • Fast procedure allows for 96 samples to be processed in 60 minutes
  • Flexible protocol accommodates sample volume inputs from 200 μL to 2 mL of blood, urine, swabs, lavage, and various transport media such as Universal Viral Transport medium, and ThinPrep and Eswab preservative fluid
  • Compatible with bronchoaveolar lavage and other respiratory samples
  • No need for carrier RNA
  • Elution volumes ranging from 50 to 100 μL


Maximum sample input in a 96-deep well or 24-deep well plate format
The reagents and enzyme mix included in the MagMAX Viral/Pathogen Ultra kit enable you to maximize the amount of sample input, thereby increasing the amount of RNA and/or DNA recovered from the most challenging pathogens. Up to 400 μL of sample can be processed per well in a KingFisher 96-deep well plate. For larger sample volumes, protocols are available to process up to 2 mL of sample per well in a KingFisher 24-deep well plate.


Validated for use with the TaqMan Microbiota array plates
Have confidence that your nucleic acid extraction kit will work with your downstream application and enjoy a complete solution from Thermo Fisher Scientific.


Nucleic acid extracted with the MagMAX Viral/Pathogen Ultra kit has been tested with the following TaqMan Microbiota assay panels:

  • Urinary tract microbiota
  • Respiratory tract microbiota
  • Vaginal microbiota


Benefits of MagMAX magnetic bead–based purification
Magnetic beads offer many benefits compared to other technologies for isolating nucleic acid. Beads bind the DNA more efficiently than glass-fiber filters, resulting in higher and more consistent yields. Additionally, because filters and vacuum manifolds are not used, there is no risk of filter clogging due to cellular particulates in samples. This clogging issue is of particular concern with protein-rich, large-volume samples such as plasma that are often used in infectious disease studies.


The Dynabeads MyOne Silane contained in the MagMAX Viral/Pathogen Ultra Nucleic Acid Isolation Kit have an optimized silica-like surface chemistry with high specific surface area that allows efficient kinetics and high sensitivity in nucleic acid capture. These aspects make Dynabeads MyOne Silane ideal for the recovery of viral RNA and DNA present at low concentrations in body fluids. Once captured, these nucleic acids can then be eluted in as little as 50 μL of elution buffer for use in downstream applications.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 55 mL Binding Solution, store at room temperature
• 100 mL Wash Solution, store at room temperature
• 10 mL Elution Solution, store at room temperature
• 1 mL Proteinase K, store at room temperature
• 2 mL DNA/RNA Binding Beads, store at room temperature,
• 5 mL Enzyme Mix, Store -5 to -30°C

Format 96 Deep-well Plate, 24 Deep-well Plate
Purification Time 60 min.
Product Line MagMAX
Quantity 100 preps
Shipping Condition Approved for shipment at Room Temperature or Dry Ice
I am getting MagMAX bead carryover in some wells of my plate. What might be causing this and how can I resolve it?

Some bead carryover is common and has not been documented to affect downstream assays. The most common cause of bead carryover is too much input sample. The best way to determine if the sample amount is the cause of the issue is to run PBS with Xeno (artificial DNA/RNA) in the lysis buffer and then check if there is still bead carryover. If that is not the cause, the initial sample may require special processing, such as bead beating or liquification. If there is still bead carryover without the original sample, and in particular, if the carryover is present in the same wells for multiple runs, then this may be an instrument alignment issue.

Do I need any additional consumables when using MagMAX kits on the MagMAX Express-96 instrument or a KingFisher instrument?

Plastic consumables need to be purchased separately in order to use the MagMAX reagents on the MagMAX Express-96 or a KingFisher instrument. The particular consumables and the amount required will vary by kit and protocol so please check the user guide for the particular MagMAX kit to be sure that all required plastic consumables are available prior to sample processing.

Where can I find Bindlt script protocol (.bdz) files for use of MagMAX kits with the KingFisher Flex Purification System and KingFisher Duo Prime Purification System?

The best place to download KingFisher Flex and KingFisher Duo Prime protocols for a particular MagMAX kit is the relevant MagMAX kit product page. The Bindlt script protocol (.bdz) files can be found under the Documents>Product literature section.

Can I scale up the MagMAX kit reaction volumes? If so, how should I do this?

Yes, reaction volumes for MagMAX kits can be scaled up. In general, the amount of MagMAX reagents should be scaled up proportionally for the increase in sample input amount. Please see the specific kit user guide for any recommendation. If none are included, then optimization may be necessary.

Will addition of BME (2-mercaptoethanol) to the MagMAX lysis buffer cause any issues with nucleic acid extraction?

Addition of BME (2-mercaptoethanol) to the lysis buffer of MagMAX kits is fine. This can be beneficial with more RNase-rich samples like certain types of tissues.
In general, we recommend adding 0.3 µL BME per 100 µL of lysate.

Which MagMAX kit do you recommend for extracting viral/bacterial RNA from swabs?

We recommend using the MagMAX Viral/Pathogen Nucleic Acid Isolation Kit (Cat. No. A42352) for extracting total nucleic acid from easily lysed viruses and Gram-negative bacteria. For difficult-to-lyse samples like Gram-positive bacteria and fungi, we recommend using the MagMAX Viral/Pathogen Ultra Kit (Cat. No. A42356).

How long is the protocol for the MagMAX Viral/Pathogen Ultra Nucleic Acid Isolation Kit?

It takes <60 min on instrument time from sample to extracted nucleic acid for up to 96 samples.

What is the lower limit for viral titer with the MagMAX Viral/Pathogen Ultra Nucleic Acid Isolation Kit?

With the MagMAX Viral/Pathogen Ultra Nucleic Acid Isolation Kit, nucleic acid has been successfully detected from as low as 50 viral copies in the starting sample.

What is the lower limit for microbes with the MagMAX Viral/Pathogen Ultra Nucleic Acid Isolation Kit?

With the MagMAX Viral/Pathogen Ultra Nucleic Acid Isolation Kit, nucleic acid has been successfully detected from as low as 1000 cfu of bacteria and yeast/fungi in the starting sample.

What types of pathogens have been tested with the MagMAX Viral/Pathogen Ultra Nucleic Acid Isolation Kit?

Multiple targets each of (-)ssRNA virus, (+)ssRNA virus, ssDNA virus, dsDNA virus, Gram negative bacteria, Gram positive bacteria, yeast, yeast-like fungi, and fungi have been tested with the MagMAX Viral/Pathogen Ultra Nucleic Acid Isolation Kit.

What sample types are compatible with the MagMAX Viral/Pathogen Ultra Nucleic Acid Isolation Kit?

Human biofluid samples (e.g., blood, serum, plasma, urine, CSF, lavage, saliva), samples stored in transport media (e.g., BD Universal Viral Transport (UVT) Media), and bacteriophage culture are compatible with the MagMAX Viral/Pathogen Ultra Nucleic Acid Isolation Kit.

What types of downstream analysis are compatible with the MagMAX Viral/Pathogen Nucleic Acid Isolation Kit and MagMAX Viral/Pathogen Ultra Nucleic Acid Isolation Kit?

The following downstream analysis are compatible with the MagMAX Viral/Pathogen Nucleic Acid Isolation Kit and MagMAX Viral/Pathogen Ultra Nucleic Acid Isolation Kit: PCR, qPCR, qRT-PCR, OpenArray analysis, microarray analysis, and next-generation sequencing.

Are the MagMAX Viral/Pathogen and MagMAX Viral/Pathogen Ultra nucleic acid isolation kits automation-compatible?

The MagMAX Viral/Pathogen and MagMAX Viral/Pathogen Ultra nucleic acid isolation kits are compatible with all KingFisher instruments (Flex, Duo Prime, and Presto) and can also be used for manual processing in 96-well plates.

What is the volume input range for the MagMAX Viral/Pathogen Nucleic Acid Isolation Kit (Cat. No. A42352) and MagMAX Viral/Pathogen Ultra Nucleic Acid Isolation Kit (Cat. No. A42356)?

For the 96-deep well plate format: 200-400 µL sample volume input (200 µL for whole blood
For the 24-deep well plate format: 500-2,000 µL sample volume input (1,000 µL for whole blood)


compliance-icons compliance-icons compliance-icons
Product Identifier
  • MagMAX Viral/Pathogen Ultra Nucleic Acid Isolation Kit
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 C
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: lysozyme, proteonase, thiocyanic acid, acrylamide

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.