Learn More
Invitrogen™ MagMAX™ Wastewater Ultra Nucleic Acid Isolation Kit with Virus Enrichment
Shop All Applied Biosystems ProductsDescription
Includes
Dynabeads Wastewater Virus Enrichment
Features of the MagMAX Wastewater Ultra Nucleic Acid Isolation Kit include:
- Automation-ready concentration and extraction protocols designed for KingFisher Flex, KingFisher APEX, and KingFisher Duo systems
- Fast procedure allows for virus enrichment and nucleic acid extraction for 24 wastewater samples in less than two hours
- Dynabeads wastewater virus enrichment is used to concentrate wastewater before extraction
- Elution volumes ranging from 50 to 100 μL
Wastewater and sewage surveillance is a useful tool for monitoring the regional spread of viral and bacterial pathogens. Surveillance at different levels of operation, such as a university dormitory or an entire city, has been shown to be a reliable way to detect the SARS-CoV-2 virus before it becomes widespread in a population. Wastewater surveillance can be used to monitor other viral targets such as influenza and norovirus. However, wastewater and sewage samples are some of the most challenging when it comes to extraction of RNA and DNA, until now.
Automated virus enrichment
Up to 10 mL of wastewater can be processed, with virus concentration followed by extraction. When the virus enrichment and extraction process is performed using a KingFisher Flex or APEX system, 24 samples can be processed in less than two hours. When the protocol is completed, pure RNA and DNA is concentrated in 50–100 μL of elution buffer. If other concentration methods, such as ultracentrifugation or precipitation are desired, the MagMAX Wastewater Ultra Nucleic Acid Isolation Kit is available for 200 μL to 1 mL input volumes and includes only extraction reagents, enabling users to use a concentration method of choice or non-concentrated wastewater samples.
Benefits of MagMAX magnetic bead–based purification
Magnetic beads offer many benefits compared to other technologies for isolating nucleic acid. Beads bind the DNA and RNA more efficiently than glass-fiber filters, resulting in higher and more consistent yields. Additionally, because filters and vacuum manifolds are not used, there is no risk of filter clogging due to cellular particulates in samples.
The Dynabeads MyOne Silane contained in the MagMAX Wastewater Ultra Nucleic Acid Isolation Kit have an optimized silica-like surface chemistry with high specific surface area that allows efficient kinetics and high sensitivity in nucleic acid capture. These aspects make Dynabeads MyOne Silane ideal for the recovery of RNA and DNA. Once captured, these nucleic acids can then be eluted in of 50–100 μL of elution buffer for use in downstream applications.
Specifications
Specifications
| Content And Storage | • 10 mL Dynabeads Wastewater Virus Enrichment; 4°C • 80 mL Lysis Buffer; room temperature • 50 mL Binding Solution; room temperature • 200 mL Wash Buffer; room temperature • 25 mL Elution Solution; room temperature • 4 mL Proteinase K; room temperature • 2 mL DNA/RNA Binding Beads; room temperature |
| Isolation Technology | Magnetic Bead |
| Sample Type | Wastewater |
| Elution Volume | 100 to 500 μL |
| Final Product Type | RNA and DNA |
| For Use With (Equipment) | KingFisher™ Duo Prime, KingFisher™ Flex, KingFisher™ Apex |
| For Use With (Application) | RT-PCR, qPCR, cDNA library construction, NGS, microarray analysis, blot hybridization, Northern/Southern blotting, in vitro translation, nuclease protection assays, nucleic acid labeling, hybridization |
| Quantity | 100 Preps |
| Shipping Condition | Room Temperature |
| Test Time | Up to 45 min. when using automated protocol |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.