missing translation for 'onlineSavingsMsg'
Learn More

Magnesium acetate, 1M aq. soln.

Product Code. 15432889
Click to view available options
Quantity:
50 mL
100 mL
Unit Size:
100mL
50mL
This item is not returnable. View return policy

Product Code. 15432889

Brand: Thermo Scientific Alfa Aesar J60041.AE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 142-72-3 | C4H6MgO4 | 142.393 g/mol

Magnesium acetate is used in molecular biology applications such as batch in vitro transcription of RNA and the crystallization of transcription factor:DNA complexes and proteins via the sitting-drop vapor-diffusion method. Also used as a buffer and to detect sodium and as a catalyst for sulfuric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium acetate is used in molecular biology applications such as batch in vitro transcription of RNA and the crystallization of transcription factor:DNA complexes and proteins via the sitting-drop vapor-diffusion method. Also used as a buffer and to detect sodium and as a catalyst for sulfuric acid.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-72-3
Molecular Formula C4H6MgO4
Molecular Weight (g/mol) 142.393
MDL Number MFCD00003539
InChI Key UEGPKNKPLBYCNK-UHFFFAOYSA-L
Synonym magnesium acetate, magnesium diacetate, cromosan, acetic acid, magnesium salt, mg acetate, magnesium di acetate, nu-mag, unii-0e95jzy48k, ccris 7883, acetic acid, magnesium salt 2:1
PubChem CID 8896
ChEBI CHEBI:62964
IUPAC Name magnesium;diacetate
SMILES CC(=O)[O-].CC(=O)[O-].[Mg+2]

Specifications

Physical Form Liquid
Quantity 100 mL
Linear Formula Mg(CH3COO)2
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 142.39
Concentration or Composition (by Analyte or Components) 1M aq. soln.
Chemical Name or Material Magnesium acetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.