missing translation for 'onlineSavingsMsg'
Learn More

Magnesium acetate tetrahydrate, 97.5%, extra pure, crystalline

Product Code. 10444195 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10444195 5 kg Plastic drum 5kg
10002730 250 g Plastic bottle 250g
10388390 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10444195 Supplier Thermo Scientific Acros Supplier No. 211570051
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 16674-78-5 | C4H14MgO8 | 214.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16674-78-5
Molecular Formula C4H14MgO8
Molecular Weight (g/mol) 214.45
MDL Number MFCD00149214
InChI Key XKPKPGCRSHFTKM-UHFFFAOYSA-L
Synonym magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p
PubChem CID 134717
SMILES O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 80.0°C
Color White
Physical Form Crystalline Powder
Quantity 5 kg
Assay Percent Range 97.5%
Linear Formula (CH3CO2)2Mg·4H2O
Fieser 15,196
Merck Index 15, 5718
IUPAC Name magnesium(2+) diacetate tetrahydrate
Formula Weight 214.46
Percent Purity 97.5%
Grade Extra Pure
Infrared Spectrum Authentic
Packaging Plastic drum
Chemical Name or Material Magnesium acetate tetrahydrate, Extra Pure
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.