Learn More
Magnesium acetate tetrahydrate, 98% (metals basis)
Description
Magnesium acetate tetrahydrate is commonly used as a source of magnesium in biological reactions It is an essential compound for a number of biochemical processes. Involved in glycolysis and the Krebs cycle, formation of cyclic-AMP and ATP.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 80°C |
| Physical Form | Crystalline |
| Quantity | 250 g |
| Beilstein | 3730605 |
| Merck Index | 14,5654 |
| Solubility Information | Freely soluble in water, alcohol |
| Formula Weight | 214.45 (142.39 Anhydrous) |
| Percent Purity | 98% |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.