missing translation for 'onlineSavingsMsg'
Learn More

Magnesium acetate tetrahydrate, 99.5%, for analysis

Product Code. p-44374 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic Bottle
Plastic drum
Unit Size:
1kg
5g
5kg
This item is not returnable. View return policy

Product Code. 10399050

Brand: Thermo Scientific Acros 270500050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16674-78-5 | C4H14MgO8 | 214.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16674-78-5
Molecular Formula C4H14MgO8
Molecular Weight (g/mol) 214.45
MDL Number MFCD00149214
InChI Key XKPKPGCRSHFTKM-UHFFFAOYSA-L
Synonym magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p
PubChem CID 134717
SMILES O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 80.0°C
Color White
pH 8 to 9 (5% soln. at 20°C)
Physical Form Crystalline Powder
Quantity 5 g
Assay Percent Range 99.5% min. (Complexometry)
Linear Formula (CH3CO2)2Mg·4H2O
Fieser 15,196
Merck Index 15,5718
Solubility Information Solubility in water: 1200g/L (15°C). Other solubilities: soluble in alcohol
IUPAC Name magnesium(2+) diacetate tetrahydrate
Formula Weight 214.46
Percent Purity 99.5%
Grade Analytical
Infrared Spectrum Authentic
Packaging Glass Bottle
Chemical Name or Material Magnesium acetate tetrahydrate, p.a.
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.