missing translation for 'onlineSavingsMsg'
Learn More

Magnesium acetate tetrahydrate, ACS, 98.0-102.0%

Product Code. 11373448
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 11373448

Brand: Thermo Scientific Alfa Aesar 036358.A4

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16674-78-5 | C4H14MgO8 | 214.45 g/mol

Used as a source of magnesium in biological reactions and as a source of acetate ion in chemical transformations. Acts as an emission control agent in coal combustion process to reduce acid rain. It is involved in glass formation process via the thermal decomposition of solids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16674-78-5
Molecular Formula C4H14MgO8
Molecular Weight (g/mol) 214.45
MDL Number MFCD00149214
InChI Key XKPKPGCRSHFTKM-UHFFFAOYSA-L
Synonym magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p
PubChem CID 134717
IUPAC Name magnesium(2+) diacetate tetrahydrate
SMILES O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 80°C
Color White
Physical Form Crystals
Quantity 2.5 kg
Assay Percent Range 98.0-102.0%
Beilstein 3730605
Solubility Information Soluble in water.
Formula Weight 214.45 (142.39anhydrous)
Grade ACS
Odor Odorless
Sensitivity Hygroscopic
Appearance White crystals
Density 1.454
Chemical Name or Material Magnesium acetate tetrahydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.