missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Acetate Tetrahydrate, Certified AR for Analysis, Fisher Chemical

Product Code. 10795331 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10795331 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10795331 Supplier Fisher Chemical Supplier No. M/0350/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 16674-78-5
Melting Point 80°C
Color White
pH 7 to 9
Packaging Plastic powder jar
Molecular Formula C4H14MgO8
MDL Number MFCD00149214
Quantity 500 g
Synonym magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p
InChI Key XKPKPGCRSHFTKM-UHFFFAOYSA-L
SMILES O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
IUPAC Name magnesium;diacetate;tetrahydrate
Molecular Weight (g/mol) 214.45
PubChem CID 134717
Formula Weight 214.398g/mol
Physical Form Solid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.