missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Acetate Tetrahydrate, Solstice

Product Code. 15662270 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15662270 500 g 500g
15642270 100 g 100g
15652270 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15662270 Supplier Honeywell-Fluka Supplier No. 228648500G
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

ACS Reagent,≥98%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16674-78-5
Molecular Formula C4H14MgO8
Molecular Weight (g/mol) 214.45
MDL Number MFCD00149214
InChI Key XKPKPGCRSHFTKM-UHFFFAOYSA-L
Synonym magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p
PubChem CID 134717
IUPAC Name magnesium;diacetate;tetrahydrate
SMILES O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O

Specifications

Quantity 500 g
Linear Formula (CH3COO)2Mg · 4H2O
UN Number NONH for all modes of transport
Beilstein 3730605
Formula Weight 214.45g/mol
Percent Purity ≥98%
Grade ACS Reagent
Chemical Name or Material Magnesium Acetate Tetrahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.