missing translation for 'onlineSavingsMsg'
Learn More

Magnesium bromide diethyl etherate, 98%

Product Code. 15409029
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15409029 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15409029 Supplier Thermo Scientific Alfa Aesar Supplier No. L15481.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 29858-07-9 | C4H10Br2MgO | 258.24 g/mol

The magnesium bromide diethyl etherate (MgBr2· OEt2)-methyl sulfide (Me2S) system is useful for the mild and chemo-selective deprotection of p-methoxybenzyl (PMB) ether. Lewis acid for catalysis of Diels-Alder reactions of homochiral 2-sulfinylmaleates with cyclopentadiene. Magnesium bromide diethyl etherate is used in the synthesis of silicon-containing azole derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The magnesium bromide diethyl etherate (MgBr2· OEt2)-methyl sulfide (Me2S) system is useful for the mild and chemo-selective deprotection of p-methoxybenzyl (PMB) ether. Lewis acid for catalysis of Diels-Alder reactions of homochiral 2-sulfinylmaleates with cyclopentadiene. Magnesium bromide diethyl etherate is used in the synthesis of silicon-containing azole derivatives.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture.
TRUSTED_SUSTAINABILITY

Specifications

CAS 29858-07-9
Flash Point 35°C (95°F)
Molecular Formula C4H10Br2MgO
Linear Formula (CH3CH2)2O·MgBr2
MDL Number MFCD00064500
Quantity 25 g
UN Number UN1325
Sensitivity Moisture sensitive
Synonym magnesium bromide diethyl etherate, magnesium bromide etherate, magnesium bromide ethyl etherate, grignard reagent
Solubility Information Hydrolyzes in water.
InChI Key JGZKUKYUQJUUNE-UHFFFAOYSA-L
SMILES Br[Mg]Br.CCOCC
IUPAC Name magnesium;ethoxyethane;dibromide
Molecular Weight (g/mol) 258.24
PubChem CID 11010650
Formula Weight 258.24
Percent Purity 98%
Chemical Name or Material Magnesium bromide diethyl etherate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.