missing translation for 'onlineSavingsMsg'
Learn More

Magnesium bromide ethyl etherate, 99%

Product Code. 10616213 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10616213 25 g Glass bottle 25g
10464833 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10616213 Supplier Thermo Scientific Acros Supplier No. 209700250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 29858-07-9 | C4H10Br2MgO | 258.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29858-07-9
Molecular Formula C4H10Br2MgO
Molecular Weight (g/mol) 258.24
MDL Number MFCD00064500
InChI Key JGZKUKYUQJUUNE-UHFFFAOYSA-L
Synonym magnesium bromide diethyl etherate, magnesium bromide etherate, magnesium bromide ethyl etherate, grignard reagent
PubChem CID 11010650
IUPAC Name magnesium;ethoxyethane;dibromide
SMILES Br[Mg]Br.CCOCC

Specifications

Color Beige to White
Flash Point 35°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula (C2H5)2O·MgBr2
Quantity 25 g
Fieser 01,629; 07,218; 13,259; 17,174
Solubility Information Solubility in water: soluble
Formula Weight 258.24
Percent Purity 99%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material Magnesium bromide ethyl etherate
Show More Show Less
compliance-icons
Product Identifier
  • Magnesium bromide ethyl etherate
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.