missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Bromide, Extra Pure, SLR, Fisher Chemical

Product Code. 10715341 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10715341 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10715341 Supplier Fisher Chemical Supplier No. M/0400/53
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 165°C
CAS 13446-53-2
Color White
pH 6.5 to 8.0
Physical Form Powder/Solid
Quantity 500 g
Molecular Formula Br2H12MgO6
MDL Number MFCD00149780
Synonym acmc-1bp1b, magnesium bromide-water 1/2/6, magnesium 2+ ion hexahydrate dibromide, magnesium 2+ hexahydrate dibromide
InChI Key LGLXXNHIGIJYQQ-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.Br[Mg]Br
IUPAC Name dibromomagnesium hexahydrate
Molecular Weight (g/mol) 292.20
PubChem CID 17749097
Formula Weight 292.21g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons
Product Identifier
  • Magnesium bromide hexahydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.