missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Bromide, TRC

Product Code. 30475491
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30475491 250 mg 250mg
30473432 5 g 5g
30471840 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30475491 Supplier Toronto Research Chemicals Supplier No. TRCM110385250MG
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-48-2
Molecular Formula Br2Mg
Molecular Weight (g/mol) 184.11
Synonym Magnesium Dibromide
IUPAC Name magnesium;dibromide
SMILES [Mg+2].[Br-].[Br-]

Specifications

Quantity 250 mg
InChI Formula InChI=1S/2BrH.Mg/h2*1H;/q;;+2/p-2
Formula Weight 181.8217
Chemical Name or Material Magnesium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.