missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Carbonate Basic, Solstice

Product Code. 15681240 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15681240 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15681240 Supplier Honeywell-Fluka Supplier No. 131171KG
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, Ph. Eur., E504, light, powder (light), 40-45% Mg (as MgO) basis

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 39409-82-0
Physical Form Powder
Quantity 1 kg
Molecular Formula C4H10Mg5O18
Linear Formula MgCO3
MDL Number MFCD00149783
UN Number NONH for all modes of transport
InChI Key YMKSTRLFWJQKFP-UHFFFAOYSA-D
SMILES O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
IUPAC Name pentamagnesium(2+) tetrahydrate dihydroxide tetracarbonate
Molecular Weight (g/mol) 467.63
Formula Weight 84.31g/mol
Grade puriss.
Concentration or Composition (by Analyte or Components) 40 to 45% Mg (as MgO) basis
Chemical Name or Material Magnesium Carbonate Basic, Meets analytical specification of Ph. Eur.BP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.