missing translation for 'onlineSavingsMsg'
Learn More

Magnesium carbonate basic, light, for analysis, 40.0-45.0% Magnesium oxide

Product Code. 10246410 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Plastic drum
Unit Size:
1kg
2.5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10246410 1 kg Plastic drum 1kg
10403705 2.5 kg Plastic drum 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10246410 Supplier Thermo Scientific Acros Supplier No. 211070010
 Restricted Item
Estimated Shipment: 20-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 39409-82-0 | C4H10Mg5O18 | 467.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39409-82-0
Molecular Formula C4H10Mg5O18
Molecular Weight (g/mol) 467.63
MDL Number MFCD00149783
InChI Key YMKSTRLFWJQKFP-UHFFFAOYSA-D
IUPAC Name pentamagnesium(2+) tetrahydrate dihydroxide tetracarbonate
SMILES O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 1 kg
Linear Formula Mg(OH)2·5H2O
Merck Index 15, 5724
Solubility Information Solubility in water: practically insoluble
Formula Weight 121.41
Percent Purity 40.0 to 45.0% (MgO)
Grade Analytical
Packaging Plastic drum
Specific Gravity 2.16
Density 2.1600g/mL
Chemical Name or Material Magnesium carbonate basic, light
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.