missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Magnesium carbonate hydroxide pentahydrate, for biochemistry, 40.0-43.5% Magnesium oxide

Product Code. 12695137 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12695137 500 g 500g
12604175 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12695137 Supplier Thermo Scientific Chemicals Supplier No. 448045000
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 56378-72-4 | C4H12Mg5O19 | 485.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56378-72-4
Molecular Formula C4H12Mg5O19
Molecular Weight (g/mol) 485.65
MDL Number MFCD00149784
InChI Key SXKPPTNZIUYFJR-UHFFFAOYSA-D
Synonym acmc-20akln, ksc492i1t, magnesium carbonate hydroxide pentahydrate, light, magnesium hydrogen carbonate hydroxide-water 1/1/1/5
IUPAC Name pentamagnesium(2+) pentahydrate dihydroxide tetracarbonate
SMILES O.O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Melting Point 350.0°C
Quantity 500 g
Linear Formula Mg(OH)2·5H2O
Formula Weight 485.69
Percent Purity 40.0 to 43.5% (Magnesium o)
Grade Biochemical
Chemical Name or Material Magnesium carbonate hydroxide pentahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.