missing translation for 'onlineSavingsMsg'
Learn More

Magnesium carbonate hydroxide tetrahydrate, White powder, MgO approx.40-43.5%

Product Code. 11367658
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367658 100 g 100g
11357658 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367658 Supplier Thermo Scientific Alfa Aesar Supplier No. 033333.22
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 39409-82-0 | C4H10Mg5O18 | 467.63 g/mol

Magnesium carbonate hydroxide tetrahydrate is used as a mixture with huntite and as filler in polymers. It acts as a flame retardant and fire retardant additive for polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39409-82-0
Molecular Formula C4H10Mg5O18
Molecular Weight (g/mol) 467.63
MDL Number MFCD00149783
InChI Key YMKSTRLFWJQKFP-UHFFFAOYSA-D
Synonym Magnesium carbonate, basic, Magnesium hydroxide carbonate
IUPAC Name pentamagnesium(2+) tetrahydrate dihydroxide tetracarbonate
SMILES O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range MgO approx.40-43.5%
Solubility Information Slightly soluble in water.
Formula Weight 467.67 (395.58 Anhydrous)
Grade Reagent
Odor Odorless
Chemical Name or Material Magnesium carbonate hydroxide tetrahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.