missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium chloride, 99.9%, (trace metal basis), pure, anhydrous
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
Packaging:
Plastic bottle
Unit Size:
10g
50g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
Specifications
Specifications
| Boiling Point | 1412.0°C |
| Melting Point | 708.0°C |
| Color | White |
| Physical Form | Beads |
| Quantity | 10 g |
| Assay Percent Range | 99.9% |
| Linear Formula | MgCl2 |
| Fieser | 11,310 |
| Merck Index | 15, 5726 |
| Solubility Information | Solubility in water: 540g/L (20°C) |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto