missing translation for 'onlineSavingsMsg'
Learn More

Melford Magnesium Chloride, Hexahydrate

Product Code. 30018480
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
Product Code. Quantity unitSize
30018480 1 kg 1kg
30018479 100 g 100g
30018481 500 g 500g
3 options
This item is not returnable. View return policy

Product Code. 30018480

Brand: Melford M240001000.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Widely used reagent in chemistry and molecular biology as a source of magnesium ions.

Widely used reagent in chemistry and molecular biology as a source of magnesium ions.

Specifications

Chemical Name or Material Magnesium Chloride, Hexahydrate
CAS 7791-18-6
Quantity 1 kg
Molecular Formula MgCl2-6H2O
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.