missing translation for 'onlineSavingsMsg'
Learn More

Melford Magnesium Chloride, Hexahydrate

Product Code. 30018481
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018481 500 g 500g
30018480 1 kg 1kg
30018479 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018481 Supplier Melford Supplier No. M24000500.0
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Widely used reagent in chemistry and molecular biology as a source of magnesium ions.

Widely used reagent in chemistry and molecular biology as a source of magnesium ions.

Specifications

Chemical Name or Material Magnesium Chloride, Hexahydrate
CAS 7791-18-6
Quantity 500 g
Molecular Formula MgCl2-6H2O
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.