missing translation for 'onlineSavingsMsg'
Learn More

Magnesium chloride hexahydrate, 97+%

Product Code. 11351639
Change view
Click to view available options
Quantity:
500 g
2 kg
10 kg
Tamaño de la unidad:
10kg
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity Tamaño de la unidad
11351639 10 kg 10kg
11341639 500 g 500g
11361639 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11351639 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor 012288.A9
 Restricted Item
In Stock
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 7791-18-6 | Cl2H12MgO6 | 203.30 g/mol

It is utilized in the polymerase chain reactions (PCR). It is used for dust control, ice control, and road stabilization. It is employed in a variety of applications such as fertilizers, mineral supplements for animals, waste water treatment, artificial seawater, textiles, paper, fireproofing agents, cements, and fire extinguishers. Pharmaceutical preparations are used in several medicines and topical or skin related applications. It acts as a coagulant in the preparation of tofu from soy milk.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Especificaciones

Melting Point 117°C (decomposition)
Color White
Physical Form Flakes
Quantity 10 kg
Odor Odorless
Sensitivity Hygroscopic
Appearance White flakes
Assay Percent Range >98%
Density 1.57
Solubility Information Highly soluble in water and alcohol. Slightly soluble in acetone and pyridine.
Formula Weight 203.30 (95.22anhydrous)
Chemical Name or Material Magnesium chloride hexahydrate
Mostrar más Mostrar menos

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.