missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Chloride Hexahydrate, 99.0-101.0%, (Crystalline/ACS Reagent Grade), MP Biomedicals

Product Code. 11450532 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450532 1 kg 1kg
11460532 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11450532 Supplier MP Biomedicals Supplier No. 0219142101
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7791-18-6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

Physical Form Crystalline
Quantity 1 kg
Assay Percent Range 99.0-101.0%
Molecular Formula MgCl2 · 6H2O
Formula Weight 203.3
Percent Purity 99 to 102%
Grade ACS Reagent
Chemical Name or Material Magnesium Chloride Hexahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.