missing translation for 'onlineSavingsMsg'
Learn More

Magnesium chloride hexahydrate, Cell Culture Reagent

Product Code. 15410257
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
Unit Size:
1kg
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15410257 250 g 250g
15420257 500 g 500g
15430257 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15410257 Supplier Thermo Scientific Alfa Aesar Supplier No. J62575.30
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 7791-18-6 | Cl2H12MgO6 | 203.30 g/mol

Magnesium Chloride, Hexahydrate is widely used as a source of magnesium ion in chemistry and molecular biology applications. In biological systems, magnesium is a co-factor for many enzymes including deoxyribonuclease (DNase) and various restriction enzymes. Also plays a role in cell membrane integrity, muscle cell physiology, cardiovascular and muscular activity, and nucleic acid structure. Magnesium chloride solution is a favorable choice as an elution buffer for antibody affinity column purifications; it is much milder on most antigens than acid elution, allowing reuse of the antigen column. Also an essential cofactor for the DNA polymerase in polymerase chain reaction (PCR) amplification. Used in wood fireproofing, wool carbonizing, and cotton fabric dressing; Also used in disinfectants, magnesia cements, fire extinguishers, floor sweeping compounds, and casein glue; Also used to make parchment paper and artificial leather; Has been used therapeutically as a cathartic and to treat bovine hypomagnesaemia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium Chloride, Hexahydrate is widely used as a source of magnesium ion in chemistry and molecular biology applications. In biological systems, magnesium is a co-factor for many enzymes including deoxyribonuclease (DNase) and various restriction enzymes. Also plays a role in cell membrane integrity, muscle cell physiology, cardiovascular and muscular activity, and nucleic acid structure. Magnesium chloride solution is a favorable choice as an elution buffer for antibody affinity column purifications; it is much milder on most antigens than acid elution, allowing reuse of the antigen column. Also an essential cofactor for the DNA polymerase in polymerase chain reaction (PCR) amplification. Used in wood fireproofing, wool carbonizing, and cotton fabric dressing; Also used in disinfectants, magnesia cements, fire extinguishers, floor sweeping compounds, and casein glue; Also used to make parchment paper and artificial leather; Has been used therapeutically as a cathartic and to treat bovine hypomagnesaemia.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

Color White
Physical Form Powder
Quantity 250 g
Merck Index 14,5662
Solubility Information Soluble in water at 20°C 546g/L. Soluble in and alcohol.
Formula Weight 203.30 (95.22 Anhydrous)
Grade Cell Culture Reagent
Odor Odorless
Chemical Name or Material Magnesium chloride hexahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.