missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Chloride Hexahydrate, Solstice
BIOPHARMA

Product Code. 15671260
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
6 kg
6 x 1 kg
6 x 2.5 kg
Unit Size:
1kg
2.5kg
6kg
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15671260 1 kg 1kg
15681260 2.5 kg 2.5kg
15601270 6 x 1 kg Pack of 6
15691260 6 kg 6kg
15611270 6 x 2.5 kg Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15671260 Supplier Honeywell-Fluka Supplier No. 131521KG
  • ONLINE EXCLUSIVE PRICE
    82.00 €
    74.05 € / 1kg
    Save 7.95 €
    9% Off
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, FCC, European Pharmacopoeia, E511, ≤0.0001% Al, 99-101%,

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

Physical Form Crystalline Solid
Quantity 1 kg
Linear Formula MgCl2 · 6H2O
UN Number NONH for all modes of transport
Formula Weight 203.3g/mol
Percent Purity 99 to 101%
Grade puriss.
Chemical Name or Material Magnesium Chloride Hexahydrate, Meets analytical specification of Ph. Eur.BP, FCC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.