missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Chloride Solution, Solstice

Product Code. 15656060
Change view
Click to view available options
Quantity:
1 L
Packaging:
Poly bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15656060 1 L Poly bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15656060 Supplier Honeywell-Fluka Supplier No. 630201L
  • ONLINE EXCLUSIVE PRICE
    41.31 €
    34.20 € / 1 L
    Save 7.11 €
    17% Off
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Volumetric, 1.0 M MgCl2

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point >100°C
Melting Point <0°C
CAS 7786-30-3
Color Colorless
Physical Form Liquid
Quantity 1 L
Molecular Formula Cl2Mg
Linear Formula MgCl2
MDL Number MFCD00011106
UN Number NONH for all modes of transport
Synonym magnesium chloride, magnesium chloride anhydrous, mgcl2, magnesium chloride, anhydrous, magnesium 2+ ion dichloride, chloromagnesite, magnesiumchlorid, magnesium ii chloride
Solubility Information Completely miscible in water
InChI Key TWRXJAOTZQYOKJ-UHFFFAOYSA-L
SMILES [Mg++].[Cl-].[Cl-]
IUPAC Name magnesium;dichloride
Molecular Weight (g/mol) 95.21
PubChem CID 5360315
ChEBI CHEBI:6636
Formula Weight 95.21g/mol
Odor Odorless
Packaging Poly bottle
Density 1.08g/cu.cm
Chemical Name or Material Magnesium chloride solution
Show More Show Less
Product Identifier
  • Magnesium chloride solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.