missing translation for 'onlineSavingsMsg'
Learn More

Magnesium chloride, ultra dry, 99.9% (metals basis)

Product Code. 11390058
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11390058

Brand: Thermo Scientific Alfa Aesar 042843.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7786-30-3 | Cl2Mg | 95.21 g/mol

Magnesium chloride is generally used as a Lewis acid catalyst. It is used as a flocculating agent, analytical reagent, anesthetic for cephalopods, and also in various magnesium cements, and ceramics. It is used for dust control, and road stabilization. It is used in fire extinguishers, polymerase chain reaction (PCR) and hydrogen storage. It is a precursor for magnesium metal and other magnesium compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7786-30-3
Molecular Formula Cl2Mg
Molecular Weight (g/mol) 95.21
MDL Number MFCD00011106
InChI Key TWRXJAOTZQYOKJ-UHFFFAOYSA-L
Synonym magnesium chloride, magnesium chloride anhydrous, mgcl2, magnesium chloride, anhydrous, magnesium 2+ ion dichloride, chloromagnesite, magnesiumchlorid, magnesium ii chloride
PubChem CID 5360315
ChEBI CHEBI:6636
SMILES [Mg++].[Cl-].[Cl-]

Specifications

Melting Point 714°C
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.9% (metals basis)
Solubility Information Highly soluble in water with evolution of heat. Slightly soluble in acetone and pyridine.
IUPAC Name magnesium;dichloride
Formula Weight 95.22
Sensitivity Hygroscopic
Density 2.32 g/mL
Chemical Name or Material Magnesium chloride, ultra dry

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.