missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium citrate, dibasic hydrate
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
Description
Magnesium citrate is used to clean stool from the intestines before surgery. It is also used as a laxative for relief of constipation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White to Yellow |
| Physical Form | Powder |
| Quantity | 1 kg |
| Merck Index | 14,5663 |
| Solubility Information | Slightly soluble in water. |
| IUPAC Name | magnesium;2-(carboxymethyl)-2-hydroxybutanedioate;hydrate |
| Formula Weight | 214.14 (Anhydrous) |
| Chemical Name or Material | Magnesium citrate dibasic hydrate |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction