missing translation for 'onlineSavingsMsg'
Learn More

Magnesium germanide, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 15452327
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15452327 25 g 25g
15442327 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15452327 Supplier Thermo Scientific Alfa Aesar Supplier No. 045517.14
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

Magnesium germanide is used as primary and secondary intermediate and in chemical research. It forms germanium hydrides on exposure to hydrochloric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-52-7
Molecular Formula GeMg2
Molecular Weight (g/mol) 121.24
MDL Number MFCD11044441
InChI Key MBWJVOFAOUBYNU-UHFFFAOYSA-N
Synonym dimagnesium germanide, magnesium germanide, germane-magnesium 1/2
PubChem CID 76518831
IUPAC Name germanium;magnesium
SMILES [Mg].[Mg].[Ge]

Specifications

Physical Form Pieces
Quantity 25 g
Linear Formula Mg2Ge
Solubility Information Soluble in water (0.0016g/100mL at 250C).
Formula Weight 121.22
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Magnesium germanide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.