missing translation for 'onlineSavingsMsg'
Learn More

Magnesium hexafluorosilicate hydrate

Product Code. 11309369
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
This item is not returnable. View return policy

Product Code. 11309369

Brand: Thermo Scientific Alfa Aesar 089018.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MgSiF6·xH2O

It is employed as an antiplaque agents and oral care agents. Also used for mothproofing textiles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula MgSiF6·xH2O
MDL Number MFCD00016196

Specifications

Melting Point 120°C (decomposition)
Physical Form Powder
Quantity 500 g
UN Number UN2853
Solubility Information Soluble in water.
Formula Weight 166.39 (Anhydrous)
Density 1.788 g/mL
Chemical Name or Material Magnesium hexafluorosilicate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.