missing translation for 'onlineSavingsMsg'
Learn More

Magnesium L-Aspartate Salt, TRC

Product Code. 30478188
Change view
Click to view available options
Quantity:
25 g
50 g
Unit Size:
25g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478188 50 g 50g
30481728 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30478188 Supplier Toronto Research Chemicals Supplier No. TRCM11041850G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18962-61-3
Molecular Formula C8H12MgN2O8
Molecular Weight (g/mol) 288.49
Synonym Magnesium L-Aspartic Acid Salt, (T-4)-Magnesate(2-) bis[L-Aspartato(2-)-N,O1]-dihydrogen, (T-4)-Magnesate(2-) bis[L-Aspartato(2-)-kN,kO1]-dihydrogen, Magnesium Complex L-Aspartic Acid, Asmag, Asmag 300, Magnesium L-Aspartate, Magnesium Aspartate, Magnesium Hydroaspartate
IUPAC Name magnesium;(3S)-3-amino-4-hydroxy-4-oxobutanoate
SMILES [Mg+2].N[C@@H](CC(=O)[O-])C(=O)O.N[C@@H](CC(=O)[O-])C(=O)O

Specifications

Quantity 50 g
InChI Formula InChI=1S/2C4H7NO4.Mg/c2*5-2(4(8)9)1-3(6)7;/h2*2H,1,5H2,(H,6,7)(H,8,9);/q;;+2/p-2/t2*2-;/m00./s1
Formula Weight 288.0444
Chemical Name or Material Magnesium L-Aspartate Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.