missing translation for 'onlineSavingsMsg'
Learn More

Magnesium matrix modifier solution, 99.9995%, Solstice

Product Code. 15675500
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15675500 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15675500 Supplier Fluka Supplier No. 428892100ML
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

TraceSELECT™, 2% Mg in <5 wt. % HNO3, 99.9995%

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions
TRUSTED_SUSTAINABILITY

Specifications

CAS 13446-18-9
Quantity 100 mL
Assay Percent Range 99.9995%
MDL Number MFCD00085308
UN Number UN3264
Beilstein 4948473
Synonym Magnesium Nitrate, Nitric Acid Solution, Magnesium Modifier
InChI Formula 1S/Mg
InChI Key FYYHWMGAXLPEAU-UHFFFAOYSA-N
IUPAC Name Magnesium Nitrate, Nitric Acid Solution, Magnesium Modifier
Molecular Weight (g/mol) 256.41 g/moL
Percent Purity 100.00%
Density 1.093g/mL at 25°C
Chemical Name or Material Magnesium matrix modifier solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.