missing translation for 'onlineSavingsMsg'
Learn More

Magnesium nitrate hexahydrate, 98+%, ACS reagent

Product Code. 10484121 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic Bottle
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10484121 500 g Plastic Bottle 500g
10125563 2.5 kg Plastic Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10484121 Supplier Thermo Scientific Acros Supplier No. 423885000
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13446-18-9 | H12MgN2O12 | 256.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-18-9
Molecular Formula H12MgN2O12
Molecular Weight (g/mol) 256.40
MDL Number MFCD00149779
InChI Key MFUVDXOKPBAHMC-UHFFFAOYSA-N
Synonym magnesium nitrate hexahydrate, magnesium dinitrate hexahydrate, unii-v85k20ljmk, dusicnan horecnaty czech, v85k20ljmk, nitric acid, magnesium salt, hexahydrate, magnesium ii nitrate 1:2 , hexahydrate, magnesium 2+ ion hexahydrate dinitrate, magnesium, reference standard solution, dusicnan horecnaty
PubChem CID 202877
IUPAC Name magnesium;dinitrate;hexahydrate
SMILES O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Boiling Point 330.0°C
Melting Point 89.0°C
Color White
pH 5.0 to 8.2 (5% soln. at 25°C)
Physical Form Adhering Crystalline Powder
Quantity 500 g
Assay Percent Range 98.0 to 102% (Complexometry)
Linear Formula Mg(NO3)2·6H2O
Flash Point 93.3°C
Merck Index 15, 5738
Solubility Information Solubility in water: 420g/L (20°C). Other solubilities: freely soluble in alcohol, liquid nh3
Formula Weight 256.4
Percent Purity ≥98%
Grade ACS Reagent
Packaging Plastic Bottle
Density 1.63
Chemical Name or Material Magnesium nitrate hexahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.