missing translation for 'onlineSavingsMsg'
Learn More

Magnesium nitrate hexahydrate, ACS, 98.0-102.0%

Product Code. 11377287
Change view
Click to view available options
Quantity:
50 g
500 g
2 kg
Unit Size:
2kg
500g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11377287 2 kg 2kg
11387287 50 g 50g
11367287 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 11377287 Supplier Thermo Scientific Alfa Aesar Supplier No. 011564.A3
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

As fertilizer, in manufacture of ammonium nitrate | CAS: 13446-18-9 | H12MgN2O12 | 256.40 g/mol

It is used in printing, chemical, agriculture and ceramics industries. It is also used in the detection of selenium in plasma samples, and concentration of nitric acid. Its fertilizer grade has 10.5% nitrogen and 9.4% magnesium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-18-9
Molecular Formula H12MgN2O12
Molecular Weight (g/mol) 256.40
MDL Number MFCD00149779
InChI Key MFUVDXOKPBAHMC-UHFFFAOYSA-N
Synonym magnesium nitrate hexahydrate, magnesium dinitrate hexahydrate, unii-v85k20ljmk, dusicnan horecnaty czech, v85k20ljmk, nitric acid, magnesium salt, hexahydrate, magnesium ii nitrate 1:2 , hexahydrate, magnesium 2+ ion hexahydrate dinitrate, magnesium, reference standard solution, dusicnan horecnaty
PubChem CID 202877
IUPAC Name magnesium;dinitrate;hexahydrate
SMILES O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Boiling Point 330°C (decomposition)
Melting Point 89°C
Physical Form Crystalline
Quantity 2 kg
Assay Percent Range 98.0-102.0%
Solubility Information Very soluble in water,alcohol and liquid ammonia.
Formula Weight 256.41 (148.32 Anhydrous)
Grade ACS
Sensitivity Hygroscopic
Density 1.46
Chemical Name or Material Magnesium nitrate, hexahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.