missing translation for 'onlineSavingsMsg'
Learn More

Magnesium oxide, 2 part ceramic adhesive paste

Product Code. p-6827160
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394808 500 g 500g
11304818 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11394808 Supplier Thermo Scientific Alfa Aesar Supplier No. 043135.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High Temperature Adhesive For Bonding Ceramic To Metal

High temperature adhesive for bonding ceramic to metal

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Mix ratio (w/w): 1.5:1 powder to liquid. Cure conditions: 1-4 hours at room temperature, 2 hours at 94°C. Components: Magnesium oxide, 2 part ceramic adhesive paste (L) and Magnesium oxide, 2 part ceramic adhesive paste (P)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Magnesium oxide, 2 part ceramic adhesive paste
Name Note Mix ratio (w/w): 1.5:1 powder to liquid. Cure conditions: 1-4 hours at room temperature, 2 hours at 94°C
Quantity 500 g
MDL Number MFCD00011109

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.