missing translation for 'onlineSavingsMsg'
Learn More

Magnesium oxide, 97+%, for analysis

Product Code. 10007542 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10007542 250 g Glass bottle 250g
10482835 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10007542 Supplier Thermo Scientific Acros Supplier No. 222532500
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1309-48-4 | MgO | 40.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-48-4
Molecular Formula MgO
Molecular Weight (g/mol) 40.30
MDL Number MFCD00011109
InChI Key CPLXHLVBOLITMK-UHFFFAOYSA-N
Synonym magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
PubChem CID 14792
IUPAC Name oxomagnesium
SMILES O=[Mg]

Specifications

Boiling Point 3600°C
Melting Point 2800°C
Color White
Physical Form Amorphous Powder
Quantity 250 g
Assay Percent Range 97% min. (on dry substance) (Complexometry)
Linear Formula MgO
Fieser 01,633; 02,256; 07,221; 16,200
Merck Index 15, 5741
Solubility Information Solubility in water: 6.2 mg/l (20°C), reacts generates heat and mg(oh)2. Other solubilities: soluble in diluted acids, insoluble in alcohol
Formula Weight 40.29
Percent Purity ≥97%
Grade Analytical
Loss on Ignition 3.0% max.
Packaging Glass bottle
Specific Gravity 3.58
Density 3.5800g/mL
Chemical Name or Material Magnesium oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.