missing translation for 'onlineSavingsMsg'
Learn More

Magnesium oxide, nanopowder, 99+% (metals basis)

Product Code. 11439970
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439970 500 g 500g
11419970 25 g 25g
11429970 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439970 Supplier Thermo Scientific Alfa Aesar Supplier No. 044733.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

S.A. >7.3m2/g | CAS: 1309-48-4 | MgO | 40.30 g/mol

Magnesium oxide is used as a laboratory reagent and an insulator in industrial cables. It acts as a reference material for calorimetry. It is widely used as a basic refractory material for crucibles, as a protective coating in plasma displays, as an oxide barrier in spin-tunneling devices, as an optical material, as a component of tubular construction in heating elements and also in libraries for preserving books. Magnesium oxide microspheres are used in the enrichment method of polycyclic aromatic hydrocarbons.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-48-4
Molecular Formula MgO
Molecular Weight (g/mol) 40.30
MDL Number MFCD00011109
InChI Key CPLXHLVBOLITMK-UHFFFAOYSA-N
Synonym magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
PubChem CID 14792
IUPAC Name oxomagnesium
SMILES O=[Mg]

Specifications

Boiling Point 3600°C
Melting Point 2852°C
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range >99% (Metal basis)
Solubility Information Soluble in hydrochloric acid and dilute acids. Insoluble in water and ethanol.
Formula Weight 40.3
Refractive Index 1.736
Appearance White powder
Density 3.58 g/cm3 at 20°C
Chemical Name or Material Magnesium oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.