missing translation for 'onlineSavingsMsg'
Learn More

Magnesium perchlorate, ACS reagent, desiccant

Product Code. 10338823 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10338823 100 g Plastic bottle 100g
10790862 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10338823 Supplier Thermo Scientific Acros Supplier No. 413461000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Magnesium perchlorate, Cl2MgO8, CAS Number-10034-81-8, anhydrous magnesium perchlorate, unii-7n77z541yf, perchloric acid, magnesium salt, perchlorate de magnesium french, dehydrite, magnesium perchlorate, anhydrous, magnesium perchlorate, anhydrone, hsdb 661, magnesium diperchlorate, 500g, White | CAS: 10034-81-8 | Cl2MgO8 | 223.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent, Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-81-8
Molecular Formula Cl2MgO8
Molecular Weight (g/mol) 223.21
MDL Number MFCD00011102
InChI Key MPCRDALPQLDDFX-UHFFFAOYSA-L
Synonym anhydrone, magnesium perchlorate, dehydrite, perchloric acid, magnesium salt, anhydrous magnesium perchlorate, unii-7n77z541yf, hsdb 661, magnesium diperchlorate, magnesium perchlorate, anhydrous, perchlorate de magnesium french
PubChem CID 24840
IUPAC Name magnesium;diperchlorate
SMILES [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Mg+2]

Specifications

Melting Point 250.0°C
Color White
Physical Form Crystalline Powder, Flakes, and Chunks
Quantity 100 g
Linear Formula Mg(ClO4)2
Merck Index 15, 5742
Solubility Information Solubility in water: 993g/L (25°C). Other solubilities: very soluble in alcohol
Formula Weight 223.21
Grade ACS Reagent
Loss on Drying 8% max. (190°C)
Packaging Plastic bottle
Chemical Name or Material Magnesium perchlorate, Desiccant
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Magnesium perchlorate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.