missing translation for 'onlineSavingsMsg'
Learn More

Magnesium perchlorate, ACS

Product Code. 11361738
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11361738 100 g 100g
11371738 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11361738 Supplier Thermo Scientific Alfa Aesar Supplier No. 011636.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Magnesium Perchlorate, Anhydrous, Cl2MgO8, CAS Number-10034-81-8, magnesium perchlorate, anhydrous, anhydrous magnesium perchlorate, anhydrone, magnesium perchlorate, magnesium diperchlorate, hsdb 661, dehydrite, perchlorate de magnesium french, perchloric acid, magnesium salt, unii-7n77z541yf, 500g | CAS: 10034-81-8 | Cl2MgO8 | 223.197 g/mol

Magnesium perchlorate is commonly utilized as a potent oxidizing agent. It is used as dehydrating agent in organic synthesis. It is practiced in many domains like pharma industry, pyrotechnics industry,in rechargeable batteries and it is a versatile tool in organic synthesis. It possesses photoelectronic and optical properties. Thus it can be used in photo electronic and optoelectronic devices. It is applied as a strong drying agent for gases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-81-8
Molecular Formula Cl2MgO8
Molecular Weight (g/mol) 223.197
MDL Number MFCD00011102
InChI Key MPCRDALPQLDDFX-UHFFFAOYSA-L
Synonym anhydrone, magnesium perchlorate, dehydrite, perchloric acid, magnesium salt, anhydrous magnesium perchlorate, unii-7n77z541yf, hsdb 661, magnesium diperchlorate, magnesium perchlorate, anhydrous, perchlorate de magnesium french
PubChem CID 24840
IUPAC Name magnesium;diperchlorate
SMILES [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Mg+2]

Specifications

Melting Point 251°C (decomposition)
Physical Form Granules
Quantity 100 g
Beilstein EM>5,
Solubility Information Magnesium perchlorate is soluble in water,ethanol.
Formula Weight 223.21
Grade ACS Reagent
Sensitivity Hygroscopic
Density 2.21 g/mL
Chemical Name or Material Magnesium perchlorate, anhydrous, For analysis ACS
compliance-icons compliance-icons
Product Identifier
  • Magnesium perchlorate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Magnesium perchlorate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.