missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Perchlorate, puriss p.a, ≥98.0% (calc. based on dry substance, KT), Honeywell Fluka™

Product Code. p-7120537 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Poly bottle
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15676070 50 g Poly bottle 50g
15666070 250 g Poly bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15676070 Supplier Honeywell-Fluka Supplier No. 6309550G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a., drying agent, ACS Reagent,≥98.0% (calc. based on dry substance, KT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-81-8
Molecular Formula Cl2MgO8
Molecular Weight (g/mol) 223.197
MDL Number MFCD00011102
InChI Key MPCRDALPQLDDFX-UHFFFAOYSA-L
Synonym anhydrone, magnesium perchlorate, dehydrite, perchloric acid, magnesium salt, anhydrous magnesium perchlorate, unii-7n77z541yf, hsdb 661, magnesium diperchlorate, magnesium perchlorate, anhydrous, perchlorate de magnesium french
PubChem CID 24840
IUPAC Name magnesium;diperchlorate
SMILES [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Mg+2]

Specifications

Physical Form Solid
Quantity 50 g
Linear Formula Mg(ClO4)2
UN Number UN1475
Formula Weight 223.21g/mol
Percent Purity ≥98.0% (calc. based on dry substance, KT)
Packaging Poly bottle
Chemical Name or Material Magnesium perchlorate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.