missing translation for 'onlineSavingsMsg'
Learn More

Magnesium phthalocyanine, tech. 90%

Product Code. 11338917
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338917 5 g 5g
11348917 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11338917 Supplier Thermo Scientific Alfa Aesar Supplier No. 038601.06
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1661-03-6 | C32H18MgN8O | 554.856 g/mol

Magnesium phthalocyanine is used in preparing membrane sensors for cyanide ions. Also used in preparation of membrane sensors for cyanide ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1661-03-6
Molecular Formula C32H18MgN8O
Molecular Weight (g/mol) 554.856
MDL Number MFCD00041953
InChI Key HTNHYCHFBSVBID-UHFFFAOYSA-N
Synonym magnesium phthalocyanine
PubChem CID 91872988
SMILES C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.O.[Mg+2]

Specifications

Quantity 5 g
Solubility Information Soluble in concentrated sulfuric acid.
Formula Weight 536.84
Percent Purity 90%
Grade Technical
Physical Form Powder
Chemical Name or Material Magnesium phthalocyanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.