missing translation for 'onlineSavingsMsg'
Learn More

Magnesium silicate, activated, for chromatography, 60-100 mesh

Product Code. 11480677
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11480677 250 g 250g
11470677 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11480677 Supplier Thermo Scientific Alfa Aesar Supplier No. B22490.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1343-88-0 | MgO3Si | 100.39 g/mol

Magnesium silicate, activated, is used in polyether adsorption and decolorization effect. Also used in chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium silicate, activated, is used in polyether adsorption and decolorization effect. Also used in chromatography.

Solubility
Insoluble in water.

Notes
Incompatible with strong acids and hydrogen fluoride.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1343-88-0
Molecular Formula MgO3Si
Molecular Weight (g/mol) 100.39
MDL Number MFCD00078249
InChI Key ZADYMNAVLSWLEQ-UHFFFAOYSA-N
Synonym Activated magnesium silicate

Specifications

Quantity 250 g
Linear Formula Mg2SiO4
Solubility Information Insoluble in water.
Formula Weight 140.71
Chemical Name or Material Magnesium silicate, activated, for chromatography

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.