missing translation for 'onlineSavingsMsg'
Learn More

Magnesium stearate, 3.8-5.0% Mg

Product Code. 10723241 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
1kg
25g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10723241 25 g Plastic bottle 25g
10723051 1 kg Plastic bottle 1kg
11924931 5 kg Plastic drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10723241 Supplier Thermo Scientific Acros Supplier No. 240760250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 557-04-0 | C36H70MgO4 | 591.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 557-04-0
Molecular Formula C36H70MgO4
Molecular Weight (g/mol) 591.27
InChI Key HQKMJHAJHXVSDF-UHFFFAOYSA-L
Synonym magnesium stearate, magnesium octadecanoate, magnesium distearate, synpro 90, octadecanoic acid, magnesium salt, dibasic magnesium stearate, petrac mg 20nf, ns-m salt, sm-p, stearic acid, magnesium salt
PubChem CID 11177
ChEBI CHEBI:9254
IUPAC Name magnesium;octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Mg+2]

Specifications

Melting Point 130.0°C to 140.0°C
Color White
Loss on Drying 6% (1g, 105°C) max.
Infrared Spectrum Authentic
Packaging Plastic bottle
Linear Formula Mg(C18H35O2)2
Quantity 25 g
Merck Index 15,5754
Solubility Information Solubility in water: insoluble. Other solubilities: decomposes in diluted acids,practically insoluble in alcohols and ether
Formula Weight 591.27
Percent Purity 3.8-5% Mg
Physical Form Fine Powder
Chemical Name or Material Magnesium stearate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.