missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Stearate, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, Fisher Chemical

Product Code. 10775911 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10775911 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10775911 Supplier Fisher Chemical Supplier No. M/0955/53
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 557-04-0
Melting Point 130°C
Color White
Packaging Plastic powder jar
Molecular Formula C36H70MgO4
MDL Number 36391
Quantity 500 g
Synonym magnesium stearate, magnesium octadecanoate, magnesium distearate, synpro 90, octadecanoic acid, magnesium salt, dibasic magnesium stearate, petrac mg 20nf, ns-m salt, sm-p, stearic acid, magnesium salt
InChI Key HQKMJHAJHXVSDF-UHFFFAOYSA-L
SMILES CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Mg+2]
IUPAC Name magnesium;octadecanoate
Molecular Weight (g/mol) 591.257
PubChem CID 11177
ChEBI CHEBI:9254
Formula Weight 591.27g/mol
Physical Form Solid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.