missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Stearate, Hyqual™ Vegetable Source NF-GenAR™ FCC, Macron Fine Chemicals™

Product Code. 15597994
Change view
Click to view available options
Quantity:
2.5 kg
Unit Size:
2.5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15597994 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15597994 Supplier Macron Fine Chemicals Supplier No. 225706
  • ONLINE EXCLUSIVE PRICE
    1235.40 €
    1078.12 € / 2.5kg
    Save 157.28 €
    12% Off
 Restricted Item
Estimated Shipment: 12-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
  • Food GMP Manufactured Product
  • GMP Manufactured Product
  • Meets B.P. Chemical Specifications
  • Meets ChP Chemical Specifications
  • Meets E.P. Chemical Specifications
  • Meets F.C.C. Requirements
  • Meets J.P. Chemical Specifications
  • Meets N.F. Requirements
  • CAUTION: For Manufacturing, processing or repackaging
  • Bulk Food Chemical
  • Bulk Pharmaceutical Chemical
  • No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product

Specifications

Melting Point 130°C to 140°C
CAS 557-04-0
Quantity 2.5 kg
Assay Percent Range 4.0 to 5.0%
Molecular Formula C36H70MgO4
MDL Number MFCD00036391
InChI Key HQKMJHAJHXVSDF-UHFFFAOYSA-L
SMILES [Mg++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O
IUPAC Name magnesium(2+) dioctadecanoate
Molecular Weight (g/mol) 591.26
Identification Passes Test
Loss on Drying ≤ 6.0%
Packaging Mall STAKMOR case
Density 1.03 g/cm³ (20°C)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.