missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Sulfate, 7-Hydrate, Crystal, Multi-Compendial, U.S.P., J.T. Baker™

Product Code. 15588014 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Unit Size:
2.5kg
500g
Product Code. Quantity Packaging unitSize
15588014 500 g Poly Bottle 500g
15598014 2.5 kg Poly Bottle 2.5kg
2 options
This item is not returnable. View return policy

Product Code. 15588014

Brand: Avantor J.T.Baker 250401

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 10034-99-8
Molecular Formula H14MgO11S
Molecular Weight (g/mol) 246.47
MDL Number MFCD00149785
InChI Key WRUGWIBCXHJTDG-UHFFFAOYSA-L
Synonym magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
PubChem CID 24843
ChEBI CHEBI:31795
IUPAC Name magnesium;sulfate;heptahydrate
SMILES O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

Specifications

Physical Form Crystals
Quantity 500 g
Formula Weight 246.47
Grade Multi-Compendial/USP
Packaging Poly Bottle
Specific Gravity 1L = 1.67kg
Chemical Name or Material Magnesium Sulfate, 7-Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.