missing translation for 'onlineSavingsMsg'
Learn More

Magnesium sulfate, ≥98.0% (KT), powder (very fine), Honeywell Fluka™

Product Code. 16675873
Change view
Click to view available options
25 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16675873 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16675873 Supplier Fluka Supplier No. 6313625KG
  • ONLINE EXCLUSIVE PRICE
    6915.00 €
    6712.54 € / 25kg
    Save 202.46 €
    2% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

Specifications

Melting Point 1,130°C (Decomposition)
CAS 7487-88-9
Color Colorless
CAS Max % ≤ 100%
pH 9
Quantity 25 kg
Molecular Formula MgO4S
Linear Formula MgSO4
InChI Key CSNNHWWHGAXBCP-UHFFFAOYSA-L
SMILES [Mg++].[O-]S([O-])(=O)=O
IUPAC Name magnesium(2+) sulfate
Molecular Weight (g/mol) 120.36
Grade ACS
Density 2.7 g/cm3
Chemical Name or Material Magnesium sulfate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.