missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Sulfate, ≥99.5%, ReagentPlus™, Solstice

Product Code. 15637960 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2 kg
12 kg
Unit Size:
12kg
1kg
2kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15637960 1 kg 1kg
15657960 500 g 500g
15647960 2 kg 2kg
15627960 12 kg 12kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15637960 Supplier Fluka Supplier No. M75061KG
  • ONLINE EXCLUSIVE PRICE
    255.00 €
    239.81 € / 1kg
    Save 15.19 €
    5% Off
 Restricted Item
Estimated Shipment: 22-04-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent Grade, anhydrous,≥99.5%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7487-88-9
Molecular Formula MgO4S
Molecular Weight (g/mol) 120.36
MDL Number MFCD00011110
InChI Key CSNNHWWHGAXBCP-UHFFFAOYSA-L
Synonym magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
PubChem CID 24083
ChEBI CHEBI:32599
SMILES [Mg++].[O-]S([O-])(=O)=O

Specifications

Quantity 1 kg
UN Number NONH for all modes of transport
IUPAC Name magnesium(2+) sulfate
Formula Weight 120.37g/mol
Percent Purity ≥99.5%
Vapor Pressure 0.1 mmHg (20°C)
Density Vapor Density: <0.01 (vs air)
Chemical Name or Material Magnesium sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.