missing translation for 'onlineSavingsMsg'
Learn More

Magnesium sulfate, 99%, extra pure, dried, contains 3 to 4 moles of water

Product Code. 10216400 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic drum
Unit Size:
10kg
1kg
2.5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10216400 2.5 kg Plastic Bottle 2.5kg
10256350 1 kg Plastic Bottle 1kg
10174573 10 kg Plastic drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10216400 Supplier Thermo Scientific Acros Supplier No. 196850025
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 22189-08-8 | MgO4S | 120.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22189-08-8
Molecular Formula MgO4S
Molecular Weight (g/mol) 120.36
MDL Number MFCD00149787
InChI Key CSNNHWWHGAXBCP-UHFFFAOYSA-L
Synonym magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
PubChem CID 24083
ChEBI CHEBI:32599
SMILES [Mg++].[O-]S([O-])(=O)=O

Specifications

Melting Point 1124.0°C
Color White
Physical Form Crystalline Powder
Quantity 2.5 kg
Assay Percent Range 98.5% min. (On dried substance) (Complexometry)
Linear Formula MgSO4·xH2O
Solubility Information Solubility in water: 360g/L (20°C). Other solubilities: soluble in glycerol, sparingly soluble in alcohol
IUPAC Name magnesium;sulfate
Formula Weight 120.36
Percent Purity 99%
Grade Extra Pure
Loss on Drying 26% to 32% (400 to 500 °C)
Packaging Plastic Bottle
Specific Gravity 2.66
Density 2.6600g/mL
Chemical Name or Material Magnesium sulfate, Dried, contains 3 to 4 Moles of Water
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.